Learn More
VEOLIA WTS ACID REAGENT CARTRIDGE
Catalog No. NC9272656
Encompass
Acid reagent cartridge for Sievers 900 Series. Replacement recommended after 6 months.
Catalog No. NC9272656 Supplier VEOLIA WTS Supplier No. APF 90310021
Acid reagent cartridge for Sievers 900 Series. Replacement recommended after 6 months.