Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Lange™ Acid Reagent Powder Pillows

Catalog No. 501994498 Shop All Hach Company Products
Encompass
Change view
Click to view available options
Quantity:
100/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-4498 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-199-4498 Supplier Hach Lange™ Supplier No. 104299
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

For determination of high range silica by the silicomolybdate method (Hach method 8185).

For determination of high range silica by the silicomolybdate method (Hach method 8185) Range: 1-100.0 mg/L SiO2. Also used in the iodometric method (Hach method 10100) for the determination of high range hypchlorite (50-150 g/L as Cl2). Pack of 100 powder pillows.

    Specifications

  • Method: 8185
  • Method Name: Silicomolybdate
  • Number of tests: 100
  • Parameter: Silica
  • Platform: Powder Pillows
  • Quantity: 100 /pk
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 25 mL
Quantity 100/Pk.
Type Powder Pillows
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.