Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Red 1, Thermo Scientific™

Catalog No. AC198700250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC198700250 25 g
AC198700050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC198700250 Supplier Thermo Scientific Chemicals Supplier No. 198700250
  • $107.70
    $91.65 / Each of 1

    Save $16.05

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemical Name or Material Acid Red 1
Absorbance ≥250 lambda 503 - 509 nm, 0.02 g/l water; >250 lambda 530 - 534 nm, 0.02 g/l water
CAS 3734-67-6
Infrared Spectrum Conforms
Loss on Drying ≤10 %
Packaging Glass bottle
Molecular Formula C18 H13 N3 Na2 O8 S2
Certificate of Analysis Available
MDL Number 00003954
Synonym 5-Acetamido-4-hydroxy-3-phenylazo-2,7-naphthalene-disulfonicacid, disodium salt; Amido Naphthol Red G; Azophloxine; C.I. 18050
Color Purple to Red
Solubility Information Soluble in water
Quantity 25 g
Physical Form Powder
Molecular Weight (g/mol) 509.43
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.