Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Red 73, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014547 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4547 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4547 Supplier MilliporeSigma Supelco Supplier No. 4982325MG
Add to Cart
Add to Cart

Acid Red 73 is a synthetically prepared sulfonated azo dye.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 5413-75-2
Physical Form Neat
Linear Formula C22H14N4 Na2O7S2
MDL Number MFCD00003907
Synonym Brilliant Crocein MOO; 7-Hydroxy-8-(4-phenylazophenylazo)-1,3-naphthalenedisulfonic acid disodium salt; Acid Red 73; Brilliant crocein scarlet MOO; Crocein Scarlet MOO
Type Analytical Standard
Quantity 25 mg
Formula Weight 556.48
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.