Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acid red 88 | 1658-56-6 | MFCD00004070 | 400.38 g/mol | C20H13N2NaO4S | 250 MG
SDP

Catalog No. 5000229027
Encompass_Preferred

Acid red 88 is an anionic monoazo dye provided as a biochemical reagent for laboratory staining, spectrophotometric assays, and analytical applications. The sodium salt form is water-soluble and supplied as a small research-use solid with manufacturer storage recommendations.

  • Anionic azo dye for staining and biochemical assays
  • Water-soluble sodium salt form for easy preparation of aqueous solutions
  • Molecular weight ≈ 400.38 g/mol; chemical formula C20H13N2NaO4S
  • Pack size suitable for small-scale experiments (250 mg)
  • Storage guidance: room temperature; in solvent, long-term stability at -80°C

Catalog No. 50-002-29027 Supplier Medchemexpress LLC Supplier No. HY118955250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Acid red 88 is an anionic monoazo dye provided as a biochemical reagent for laboratory staining, spectrophotometric assays, and analytical applications. The sodium salt form is water-soluble and supplied as a small research-use solid with manufacturer storage recommendations.

  • Anionic azo dye for staining and biochemical assays
  • Water-soluble sodium salt form for easy preparation of aqueous solutions
  • Molecular weight ≈ 400.38 g/mol; chemical formula C20H13N2NaO4S
  • Pack size suitable for small-scale experiments (250 mg)
  • Storage guidance: room temperature; in solvent, long-term stability at -80°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.