Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Red 92 85.0+%, TCI America™
SDP

Catalog No. T005025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T005025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T005025G Supplier TCI America Supplier No. T005025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18472-87-2
Molecular Formula C20H4Br4Cl4O5
MDL Number MFCD00070627
Synonym Phloxine B, 2′C,4′C,5′C,7′C-Tetrabromo-3,4,5,6-tetrachlorofluorescein Sodium Salt

Specifications

Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 829.63
Percent Purity ≥85.0% (E)
Chemical Name or Material Acid Red 92
EINECSNumber (5)-1514
RTECSNumber LM5900000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.