Learn More
Medchemexpress LLC Acid Red sodium | 3244-88-0 | 25 G
SDP
Catalog No. 5000414210
Encompass_Preferred
Acid Red is a class of synthetic dyes commonly used in the textile, paper, and ink industries. It falls under the category of acid dyes, meaning it is water-soluble and possesses acidic properties. This dye produces bright, vibrant colors that resist fading easily, though their durability might be less compared to some other dye types. It can be applied to various fibers, including wool, silk, and nylon, as well as paper and leather. For research purposes, Acid Red serves as a biochemical reagent, usable as a biological material or organic compound in life science-related studies.
  • Synthetic dye
  • Water-soluble
  • Acidic properties
  • Produces bright, vibrant colors
  • Colors do not fade easily
  • Can be used on wool, silk, nylon, paper, and leather
  • Biochemical reagent for life science research
Catalog No. 50-004-14210 Supplier Medchemexpress LLC Supplier No. 25GHYD0854
Acid Red is a class of synthetic dyes commonly used in the textile, paper, and ink industries. It falls under the category of acid dyes, meaning it is water-soluble and possesses acidic properties. This dye produces bright, vibrant colors that resist fading easily, though their durability might be less compared to some other dye types. It can be applied to various fibers, including wool, silk, and nylon, as well as paper and leather. For research purposes, Acid Red serves as a biochemical reagent, usable as a biological material or organic compound in life science-related studies.
  • Synthetic dye
  • Water-soluble
  • Acidic properties
  • Produces bright, vibrant colors
  • Colors do not fade easily
  • Can be used on wool, silk, nylon, paper, and leather
  • Biochemical reagent for life science research