Learn More
MSC Scot's Tuff pH Neutralizing Carpet Rinse, Concentrated, 4 gal
Catalog No. 502846093
Encompass
This product is an acid rinse for carpet and upholstery, designed to neutralize alkaline residues and restore pH balance. It is concentrated and sold in a pack of four 1-gallon bottles.
  • Neutralizes alkaline residues
  • Restores pH balance to carpets and upholstery
  • Concentrated formula
  • Suitable for professional use
  • Supplied in convenient 1-gallon bottles
Catalog No. 50-284-6093 Supplier MSC Supplier No. 36152767
This product is an acid rinse for carpet and upholstery, designed to neutralize alkaline residues and restore pH balance. It is concentrated and sold in a pack of four 1-gallon bottles.
  • Neutralizes alkaline residues
  • Restores pH balance to carpets and upholstery
  • Concentrated formula
  • Suitable for professional use
  • Supplied in convenient 1-gallon bottles