Learn More
Grainger Justrite Acid Safety Cabinet, Wood Laminate, 32 Gal., 1 Ea.
Catalog No. 502792487
Encompass_Preferred
This acid safety cabinet is engineered for the secure storage of highly corrosive liquids, including phenol, nitric acid, and sulfuric acid. Featuring robust wood laminate construction and stainless steel hardware, this cabinet provides excellent resistance to harsh chemical environments. It includes an adjustable shelving system to accommodate diverse container sizes and is designed for use in laboratory and industrial settings to ensure safe chemical segregation.
  • Provides 121 L (32 gal.) capacity for up to 49, 2.5 L bottles
  • Constructed from durable wood laminate with stainless steel handles and hinges
  • Features two shelves adjustable on 32 mm (1-1/4 in.) centers
  • Equipped with reflective warning labels for increased visibility
  • Includes adjustable leg levelers and a double key set for enhanced security
Catalog No. 50-279-2487 Supplier Grainger Supplier No. 1AG24
This acid safety cabinet is engineered for the secure storage of highly corrosive liquids, including phenol, nitric acid, and sulfuric acid. Featuring robust wood laminate construction and stainless steel hardware, this cabinet provides excellent resistance to harsh chemical environments. It includes an adjustable shelving system to accommodate diverse container sizes and is designed for use in laboratory and industrial settings to ensure safe chemical segregation.
  • Provides 121 L (32 gal.) capacity for up to 49, 2.5 L bottles
  • Constructed from durable wood laminate with stainless steel handles and hinges
  • Features two shelves adjustable on 32 mm (1-1/4 in.) centers
  • Equipped with reflective warning labels for increased visibility
  • Includes adjustable leg levelers and a double key set for enhanced security