Learn More
Echelon Biosciences Research Labs Acid Sphingomyelinase Activity Assay Kit
SDP
Supplier:  Echelon Biosciences Research Labs K3200EAKIT
Encompass
The Acid Sphingomyelinase Assay Kit is an enzyme assay that measures acid sphingomyelinase activity in biological samples through the direct hydrolysis of a fluorogenic substrate.Assay sensitivity: 3.125 pmol/hourSample Type: tissue homogenates, cell lysatesSample Volume: 20 L/sample (for duplicate points) This item is non returnable
Catalog No. 50-720-446