Learn More
Creative Biomart Acid Sphingomyelinase (SMPD1) Protein
Supplier:  Creative Biomart THP0283
Encompass_Preferred
Recombinant Human Acid sphingomyelinase protein (1-364 aa) is produced by Wheat germ system. This protein could converts sphingomyelin to ceramide. Also has phospholipase C activities toward 1,2-diacylglycerolphosphocholine and 1,2-diacylglycerolphosphoglycerol. Isoform 2 and isoform 3 have lost catalytic activity.
Catalog No. 50-208-4334