Learn More
AVANTOR MASTERFLEX (VWR LLC) Hand-operated acid-transfer pump, 6 strokes/gallon, female buttress adapter
Catalog No. NC1572563
Encompass
Ideal for most acid/base or ionic salt applications
Catalog No. NC1572563 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0643340
Ideal for most acid/base or ionic salt applications