Learn More
Ops Diagnostics BAWG 4000-200-18 ACID WASH 4MM SILICA BEADS
Catalog No. NC1265514
Encompass
These large silica beads are useful for homogenizing soft tissues in harsh buffers. Beads are acid washed to remove impurities and silica dust. Silica beads are a cost effective alternative to zirconium oxide and stainless steel when high density media is unnecessary or impractical. Packaged in bottles of 200 grams.
Catalog No. NC1265514 Supplier Ops Diagnostics Supplier No. BAWG 400020018
These large silica beads are useful for homogenizing soft tissues in harsh buffers. Beads are acid washed to remove impurities and silica dust. Silica beads are a cost effective alternative to zirconium oxide and stainless steel when high density media is unnecessary or impractical. Packaged in bottles of 200 grams.