Learn More
Selleck Chemical LLC Acid Yellow 36
SDP
Catalog No. 5000002969
Encompass_Preferred
Acid Yellow 36 (Metanil Yellow) is a dye of the azo class and used as a pH indicator and it has a color change from red to yellow between pH 1 2 and 2 3
Catalog No. 50-000-02969 Supplier Selleck Chemical LLC Supplier No. S618925MG
Acid Yellow 36 (Metanil Yellow) is a dye of the azo class and used as a pH indicator and it has a color change from red to yellow between pH 1 2 and 2 3