Learn More
Medchemexpress LLC Acid Yellow 36 (Metanil Yellow) | 587-98-4 | 70.0% | 50 MG
SDP
Catalog No. 5000342616
Encompass_Preferred
Acid Yellow 36, also known as Metanil Yellow, is an azo dye that functions as a pH indicator, changing color from red at pH 1.2 to yellow at pH 2.3. It finds application in various industries including leather, paper, and textiles. Furthermore, Acid Yellow 36 acts as a bifunctional inducer of specific isozymes of P-450 and cytosolic enzymes.
  • Acts as an azo dye and pH indicator.
  • Changes color from red at pH 1.2 to yellow at pH 2.3.
  • Used in the leather, paper, and textile industries.
  • Functions as a bifunctional inducer of specific P-450 isozymes and cytosolic enzymes.
  • Induces a specific form of microsomal P-450 and cytosolic glutathione-S-transferase (GST) and quinone reductase (QR), potentially involving a cytosolic Ah receptor.
Catalog No. 50-003-42616 Supplier Medchemexpress LLC Supplier No. HY12836950MG
Add to Cart
Add to Cart
Acid Yellow 36, also known as Metanil Yellow, is an azo dye that functions as a pH indicator, changing color from red at pH 1.2 to yellow at pH 2.3. It finds application in various industries including leather, paper, and textiles. Furthermore, Acid Yellow 36 acts as a bifunctional inducer of specific isozymes of P-450 and cytosolic enzymes.
  • Acts as an azo dye and pH indicator.
  • Changes color from red at pH 1.2 to yellow at pH 2.3.
  • Used in the leather, paper, and textile industries.
  • Functions as a bifunctional inducer of specific P-450 isozymes and cytosolic enzymes.
  • Induces a specific form of microsomal P-450 and cytosolic glutathione-S-transferase (GST) and quinone reductase (QR), potentially involving a cytosolic Ah receptor.