Learn More
Description
- Chemical Name: Acid Yellow 36
- Synonym: Metanil Yellow
- Physical State (20°C): Solid
- Appearance: Light yellow to brown powder to crystal
- Purity: min. 85.0 area%
- Molecular Formula: C18H14N3NaO3S
- Molecular Weight: 375.38
- CAS RN: 587-98-4
- Absorption Maximum: 415.0 to 419.0 nm (MeOH)
- Storage Temperature: Room temperature (recommended in a cool and dark place, <15°C)
Specifications
Specifications
| Color | Pale Yellow to Greyish Yellow Red |
| Quantity | 25 g |
| Physical Form | Crystal/Powder |
| Formula Weight | 375.38 |
| Grade | Indicator |
| Chemical Name or Material | Acid Yellow 36 |
Safety and Handling
| EINECSNumber | (5)-1405 |
| RTECSNumber | DB7329500 |
| TSCA | Yes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
