Learn More
Creative Biomart Acidic Fibroblast Growth Factor (FGF-a) Protein
Catalog No. 502084283
Encompass_Preferred
Fibroblast Growth Factor-acidic Human Recombinant (FGF-1) produced in E. Coli is a single, non-glycosylated, polypeptide chain containing 140 amino acids and having a molecular mass of approximately 15.8kDa. The FGF acidic is purified by proprietary chromatographic techniques.
Catalog No. 50-208-4283 Supplier Creative Biomart Supplier No. THP0232
Fibroblast Growth Factor-acidic Human Recombinant (FGF-1) produced in E. Coli is a single, non-glycosylated, polypeptide chain containing 140 amino acids and having a molecular mass of approximately 15.8kDa. The FGF acidic is purified by proprietary chromatographic techniques.