Learn More
Bachem Ophthalmic Acid
SDP
Supplier:  Bachem 4001847.1000
Encompass_Preferred
Ophthalmic Acid, H-Glu(Abu-Gly-OH)-OH, 495-27-2, C11H19N3O6, Mr 289.29, 1g. H-g-Glu-Abu-Gly-OH. Acidic peptide of the lens. Carba analog of reduced glutathione. Ophthalmic acid could be used as a biomarker in oxidative stress.
Catalog No. 50-194-6872