Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acidic Titration Solvent, Ricca Chemical
SDP

Catalog No. R02730004A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R02730004A 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R02730004A Supplier Ricca Chemical Company Supplier No. R02730004A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Acidic Titration Solvent
CAS 6131-90-4, 64-19-7, 67-63-0, 7732-18-5
Solubility Information Solubility in Water: Infinite
Grade Laboratory
Odor Characteristic alcohol, slight acetic acid, odor
Packaging Natural Poly Bottle
Solvent or Matrix Nonaqueous Solution
Quantity 4 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.