Learn More
Innovive Inc Acidified Water - Rat - 850ml
Supplier:  Innovive Inc RWB850AS
Encompass
This item has a minimum order quantity of 360 bottles per supplier requirements.Plastic rat water bottle pre-filled with laboratory-grade acidified water (2.5-3.0 pH).
Catalog No. NC2175660