Learn More
ATCC Pfizer Pneumococcal; polysaccharide powder; Type 30 (15A) 10mg
Catalog No. 502380737
Encompass
Purified pneumococcal polysaccharide type 15A is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 15A can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-0737 Supplier ATCC Supplier No. 537X
Purified pneumococcal polysaccharide type 15A is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 15A can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.