Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acids Surrogate Standard Mix (For EPA 8270C)

Catalog No. usism290n1
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-ISM-290N-1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-ISM-290N-1 Supplier Agilent Technologies Supplier No. ISM290N1
Only null left
Add to Cart
Add to Cart

Page 312C, Designates A2LA-Certified product

Specifications

Chemical Name or Material Acids Surrogate Standard Mix
Packaging Ampule
Quantity 1 x 1 mL
Grade EPA
Concentration or Composition (by Analyte or Components) Each Component at 2000μg/mL in Methanol:
2-Fluorophenol
Phenol-d5
2, 4, 6-Tribromophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.