Learn More
Cayman Chemical AcIETDAFCtrIfluoroACTAte 10mg
SDP
Catalog No. 502966775
Encompass_Preferred
A fluorogenic substrate that can be cleaved specifically by caspase-8, caspase-3 processing enzyme, caspase-10, and granzyme B
Catalog No. 50-296-6775 Supplier Cayman Chemical Supplier No. 1748010 MG
A fluorogenic substrate that can be cleaved specifically by caspase-8, caspase-3 processing enzyme, caspase-10, and granzyme B