Learn More
Medchemexpress LLC Acifluorfen-methyl | 50594-67-7 | 99.69% | 375.68 | 1 ML
SDP
Catalog No. 5000360784
Encompass_Preferred
Acifluorfen-methyl is an inhibitor of protoporphyrinogen oxidase (Protox), an enzyme involved in heme and chlorophyll biosynthesis. It also functions as a photobleaching herbicide and is intended for research use only.
  • Inhibits protoporphyrinogen oxidase (Protox)
  • Functions as a photobleaching herbicide
  • Suitable for research applications
Catalog No. 50-003-60784 Supplier Medchemexpress LLC Supplier No. HY14736510MM/1ML
Acifluorfen-methyl is an inhibitor of protoporphyrinogen oxidase (Protox), an enzyme involved in heme and chlorophyll biosynthesis. It also functions as a photobleaching herbicide and is intended for research use only.
  • Inhibits protoporphyrinogen oxidase (Protox)
  • Functions as a photobleaching herbicide
  • Suitable for research applications