Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversey™ Acifoam™ VF10

Catalog No. 17000049
Change view
Click to view available options
Volume (Metric):
18.9 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
17-000-049 18.9 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-000-049 Supplier Diversey™ Supplier No. 57746100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Acifoam cleaner is designed for daily and periodic application in the food, beverage and dairy industries.

  • Effective scale remover
  • Versatile - Suitable for use on aluminum
  • Easy to use & Free-rinsing
  • Foam application extends contact time with vertical surfaces

Specifications

Product Type Acid Cleaner
Sterility Non-sterile
Appearance Red
Certifications/Compliance CFIA, Kosher
Container Type Pail
For Use With (Application) Descaling surfaces
Quantity 1 Ea.
Volume (English) 5 gal.
Volume (Metric) 18.9 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.