Learn More
New England Biolabs, Inc. AciI – 1000 units
SDP
Catalog No. 50812226
Encompass
A restriction endonuclease that recognizes the sequence C^CG_C.
Catalog No. 50-812-226 Supplier New England Biolabs, Inc. Supplier No. R0551L
A restriction endonuclease that recognizes the sequence C^CG_C.