Learn More
Aviva Systems Biology Corp Acin1 Antibody - N-terminal region (ARP58256_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58256_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Acin1. It was validated on Western Blot. . Alternate names: 2610036I19Rik, 2610510L13Rik, Acinus, Acn, C79325, acinusL, acinusS, mKIAA0670ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-456