Learn More
ATCC Acinetobacter baumannii
Supplier:  ATCC BAA1795
Encompass
This multidrug-resistant strain of Acinetobacter baumannii was isolated in 2008 from nasotracheal aspirate in Maryland.
Catalog No. 50-237-9805