Learn More
ATCC Acinetobacter baumannii
Catalog No. 502379807
Encompass
This strain of Acinetobacter baumannii was isolated in 2008 from a blood sample from Washington, DC. This strain is multidrug resistant and has applications in antimicrobial resistance research.
Catalog No. 50-237-9807 Supplier ATCC Supplier No. BAA1797
Add to Cart
Add to Cart
This strain of Acinetobacter baumannii was isolated in 2008 from a blood sample from Washington, DC. This strain is multidrug resistant and has applications in antimicrobial resistance research.