Learn More
ATCC Acinetobacter baumannii
Supplier:  ATCC BAA1794
Encompass
This multidrug-resistant strain of Acinetobacter baumannii was isolated in 2008 from a sputum sample in Maryland. This strain is propagated aerobically at 37°C in trypticase soy medium.
Catalog No. 50-238-1367