Learn More
ATCC Acinetobacter baumannii
Supplier:  ATCC BAA1793
Encompass
This whole-genome sequenced strain of Acinetobacter baumannii was isolated in 2008 from a sputum sample in Washington, DC. This bacterium is multidrug-resistant and can be used in antimicrobial resistance research.
Catalog No. 50-238-1365