Learn More
ATCC Acinetobacter baumannii
Supplier: ATCC BAA747
Acinetobacter baumannii strain Vitek #109234 [API 8310086, NSB 4221] is a clinical specimen isolated from ear pus. This bacterial culture can be used as a quality control strain for the VITEK 2 GN Comprehensive QC Set.