Learn More
ATCC Acinetobacter baumannii; Strain 2208; genomic DNA
Catalog No. 502383465
Encompass
Genomic DNA isolated from Acinetobacter baumannii strain 2208 (ATCC 19606). This whole-genome sequenced product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-3465 Supplier ATCC Supplier No. 19606D5
Genomic DNA isolated from Acinetobacter baumannii strain 2208 (ATCC 19606). This whole-genome sequenced product can be used in PCR and other molecular biology applications.