Learn More
ATCC Acinetobacter baylyi; Strain ADP1 (BD413)
Catalog No. 502384385
Encompass
Acinetobacter baylyi strain ADP1 (BD413) is an unencapsulated mutant derived from ATCC 33304. This whole-genome sequenced strain can be used in transformation and gene mapping.
Catalog No. 50-238-4385 Supplier ATCC Supplier No. 33305
Acinetobacter baylyi strain ADP1 (BD413) is an unencapsulated mutant derived from ATCC 33304. This whole-genome sequenced strain can be used in transformation and gene mapping.