Learn More
ATCC Acinetobacter calcoaceticus
Catalog No. 502382107
Encompass
Acinetobacter calcoaceticus strain HO-1 is a bacterial strain isolated from soil in Iowa, USA. This strain produces waxes from hydrocarbons.
Catalog No. 50-238-2107 Supplier ATCC Supplier No. 14987
Acinetobacter calcoaceticus strain HO-1 is a bacterial strain isolated from soil in Iowa, USA. This strain produces waxes from hydrocarbons.