Learn More
ATCC Acinetobacter calcoaceticus
Supplier:  ATCC 23055
Encompass
Acinetobacter calcoaceticus strain 46 [1, IFO 13718] is a whole-genome sequenced bacterial type strain that can produce isoprene.
Catalog No. 50-238-2602
 Restricted Item