Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Acinetobacter calcoaceticus Z160, Titered
SDP

Catalog No. 22158017 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-017 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-017 Supplier ZeptoMetrix Supplier No. 0804096
Add to Cart
Add to Cart

Can be used to determine limit of detection (LOD), in diagnostic assay development or crossreactivity studies

  • Each frozen aliquot contains 1mL of pure, titered culture of Acinetobacter calcoaceticus Z160
  • Identification of this organism was confirmed by 16S sequencing
  • Purity of the culture was monitored by Gram staining and additional culturing
  • Freezing medium contains 15% glycerol as cryoprotectant

Specifications

Content And Storage Should be stored at -65°C or below.
Format Titered
Product Type Titered Live Bacteria Control
Quantity 1 mL
Species Acinetobacter calcoaceticus Z160
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.