Learn More
ATCC Staphylococcus aureus; subsp. aureus
Catalog No. 502379717
Encompass
Penicillin sensitive. Does not produce penicillinase. Usually lysed by phages 7, 47, 53, 54 and 77. According to the depositor, this item is resistant to 25 μg of tetracycline, but sensitive to 50 μg.
Catalog No. 50-237-9717 Supplier ATCC Supplier No. 27217
Penicillin sensitive. Does not produce penicillinase. Usually lysed by phages 7, 47, 53, 54 and 77. According to the depositor, this item is resistant to 25 μg of tetracycline, but sensitive to 50 μg.