Learn More
ATCC Shewanella algae; Strain AmMS 201
Catalog No. 502381602
Encompass
Shewanella algae strain AmMS 201 is a whole-genome sequenced bacterium that was isolated from a clinical specimen. This strain has applications as a quality control strain for MicroScan™ products.
Catalog No. 50-238-1602 Supplier ATCC Supplier No. 49138
Shewanella algae strain AmMS 201 is a whole-genome sequenced bacterium that was isolated from a clinical specimen. This strain has applications as a quality control strain for MicroScan™ products.