Learn More
Medchemexpress LLC HY-B0283 50mg Medchemexpress, Acipimox CAS:51037-30-0 Purity:>98%
SDP
Catalog No. 502082446
Encompass_Preferred
Medchemexpress, HY-B0283 50mg Acipimox CAS:51037-30-0 Acipimox (K-9321), a nicotinic acid analogue, is an antilipolytic compound. Acipimox acutely inhibits lipolysis and suppresses systemic levels of free fatty acids (FFAs) and improves insulin sensitivity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2446 Supplier Medchemexpress LLC Supplier No. HYB028350MG
Medchemexpress, HY-B0283 50mg Acipimox CAS:51037-30-0 Acipimox (K-9321), a nicotinic acid analogue, is an antilipolytic compound. Acipimox acutely inhibits lipolysis and suppresses systemic levels of free fatty acids (FFAs) and improves insulin sensitivity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.