Learn More
Medchemexpress LLC Acipimox-d4 | 1246816-28-3 | 1 MG
SDP
Catalog No. 5000405967
Encompass_Preferred
Acipimox-d4 is a deuterium-labeled version of Acipimox (K-9321), which is a nicotinic acid analogue known for its antilipolytic properties. This compound effectively inhibits lipolysis and reduces systemic levels of free fatty acids (FFAs), thereby improving insulin sensitivity. It is intended for research use only and not for patient administration.
  • Deuterium labeled compound
  • Nicotinic acid analogue
  • Exhibits antilipolytic activity
  • Suppresses systemic free fatty acids
  • Improves insulin sensitivity
  • Suitable as a tracer in research
  • Can serve as an internal standard for quantitative analysis (e.g., NMR, GC-MS, LC-MS)
  • Incorporation of stable heavy isotopes can affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-05967 Supplier Medchemexpress LLC Supplier No. HYB0283S1MG
Acipimox-d4 is a deuterium-labeled version of Acipimox (K-9321), which is a nicotinic acid analogue known for its antilipolytic properties. This compound effectively inhibits lipolysis and reduces systemic levels of free fatty acids (FFAs), thereby improving insulin sensitivity. It is intended for research use only and not for patient administration.
  • Deuterium labeled compound
  • Nicotinic acid analogue
  • Exhibits antilipolytic activity
  • Suppresses systemic free fatty acids
  • Improves insulin sensitivity
  • Suitable as a tracer in research
  • Can serve as an internal standard for quantitative analysis (e.g., NMR, GC-MS, LC-MS)
  • Incorporation of stable heavy isotopes can affect pharmacokinetic and metabolic profiles of drugs