Learn More
Enzo Life Sciences Acitretin (10mg). CAS: 55079-83-9
SDP
Supplier:  Enzo Life Sciences BMLGR1080010
Encompass_Preferred
Acitretin is a synthetic aromatic analog of all trans retinoic acid with increased stability in vivo. It is the active free acid form of the antipsoriatic drug etretinate. Displays antiproliferative effect and induces apoptosis in Kaposi's sarcoma cells. Purity: ≥98% (TLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-0922