Learn More
Cayman Chemical AcItretIn-d3 1mg
SDP
Catalog No. 502937800
Encompass_Preferred
An internal standard for the quantification of acitretin by GC- or LC-MS
Catalog No. 50-293-7800 Supplier Cayman Chemical Supplier No. 286961 MG
Add to Cart
Add to Cart
An internal standard for the quantification of acitretin by GC- or LC-MS