Learn More
Lkt Laboratories Acivicin, ≥98%, 10MG
SDP
Supplier:  Lkt Laboratories A093410MG
Encompass_Preferred
Glutamine analog; γ-glutamyl transferase, CTP synthetase, GMP synthetase, FGAM synthetase inhibitor. CAS: 42228-92-2. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. 50-153-4861