Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acivicin, 98+%

Catalog No. AAJ63105MA
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63105MA 10 mg
AAJ63105MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63105MA Supplier Thermo Scientific Chemicals Supplier No. J63105MA
Add to Cart

CAS: 42228-92-2 | C5H7ClN2O3 | 178.572 g/mol

Inhibits glutamine amidotransferases in purine and pyrimidine synthetic pathways. Tumor growth inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Inhibits glutamine amidotransferases in purine and pyrimidine synthetic pathways
  • Tumor growth inhibitor
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42228-92-2
Molecular Formula C5H7ClN2O3
Molecular Weight (g/mol) 178.572
MDL Number MFCD00058450
InChI Key QAWIHIJWNYOLBE-OKKQSCSOSA-N
Synonym acivicin, antibiotic at 125, acivicine, acivicino, acivicinum, unii-o0x60k76i6, acia, alphas,5s-alpha-amino-3-chloro-2-isoxazoline-5-acetic acid, acivicin usan:inn, acivicine inn-french
PubChem CID 294641
ChEBI CHEBI:74545
IUPAC Name (2S)-2-amino-2-[(5S)-3-chloro-4,5-dihydro-1,2-oxazol-5-yl]acetic acid
SMILES C1C(ON=C1Cl)C(C(=O)O)N

Specifications

Melting Point 209°C to 211°C
Color White
Quantity 10 mg
UN Number UN2811
Solubility Information Soluble in water at 10mg/ml with warming
Formula Weight 178.57
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material Acivicin
missing translation for 'healthHazard1' H301
missing translation for 'healthHazard2' GHS H Statement
H301
Toxic if swallowed.
missing translation for 'healthHazard3' P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber NY2103000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.