Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acivicin hydrochloride | 161922-40-3 | MFCD32874156 | 99.1% | 215.03 | C5H8Cl2N2O3 | 1 ML
SDP

Catalog No. 5000301148
Encompass_Preferred

Acivicin hydrochloride is a Streptomyces-derived natural product that inhibits γ-glutamyl transpeptidase (GGT). It has reported anti-cancer and anti-parasitic activities and is supplied in solid forms or as a ready-to-use 10 mM solution in DMSO for research applications.

  • Acts as a γ-glutamyl transpeptidase (GGT) inhibitor.
  • Derived from Streptomyces sviceus.
  • Reported to cross the blood-brain barrier.
  • Available as solid (mg scale) and 10 mM solution in DMSO.
  • CAS number 161922-40-3.
  • Chemical formula C5H8Cl2N2O3; molecular weight 215.03.
  • Reported purity 99.12%.
  • Solubility: DMSO 100 mg/mL; in vivo formulations and ≥2.5 mg/mL indicated.
  • Storage guidance: 4°C for solids; in solvent -80°C (6 months) or -20°C (1 month) under nitrogen.

Catalog No. 50-003-01148 Supplier Medchemexpress LLC Supplier No. HYW016586A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acivicin hydrochloride is a Streptomyces-derived natural product that inhibits γ-glutamyl transpeptidase (GGT). It has reported anti-cancer and anti-parasitic activities and is supplied in solid forms or as a ready-to-use 10 mM solution in DMSO for research applications.

  • Acts as a γ-glutamyl transpeptidase (GGT) inhibitor.
  • Derived from Streptomyces sviceus.
  • Reported to cross the blood-brain barrier.
  • Available as solid (mg scale) and 10 mM solution in DMSO.
  • CAS number 161922-40-3.
  • Chemical formula C5H8Cl2N2O3; molecular weight 215.03.
  • Reported purity 99.12%.
  • Solubility: DMSO 100 mg/mL; in vivo formulations and ≥2.5 mg/mL indicated.
  • Storage guidance: 4°C for solids; in solvent -80°C (6 months) or -20°C (1 month) under nitrogen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.