Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acivicin MP Biomedicals

Catalog No. ICN15425910 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15425910 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15425910 Supplier MP Biomedicals, Inc Supplier No. 15425910

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 42228-92-2
Molecular Formula C5H7ClN2O3
Molecular Weight (g/mol) 178.572
InChI Key QAWIHIJWNYOLBE-OKKQSCSOSA-N
Synonym acivicin, antibiotic at 125, acivicine, acivicino, acivicinum, unii-o0x60k76i6, acia, alphas,5s-alpha-amino-3-chloro-2-isoxazoline-5-acetic acid, acivicin usan:inn, acivicine inn-french
PubChem CID 294641
ChEBI CHEBI:74545
IUPAC Name (2S)-2-amino-2-[(5S)-3-chloro-4,5-dihydro-1,2-oxazol-5-yl]acetic acid
SMILES C1C(ON=C1Cl)C(C(=O)O)N

Specifications

Assay Percent Range 99%
Quantity 10 mg
Formula Weight 178.6
Chemical Name or Material Acivicin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.