Learn More
Kd Medical Inc ACK LYSING 1L BOTTLE
SDP
Catalog No. 501019080
Encompass_Preferred
Ammonium-Chloride-Potassium is used to lyse red blood cells in preparations containing white blood cells. One 1 Liter bottle.
Catalog No. 50-101-9080 Supplier Kd Medical Inc Supplier No. RGF3015
Ammonium-Chloride-Potassium is used to lyse red blood cells in preparations containing white blood cells. One 1 Liter bottle.