Learn More
Kd Medical Inc ACK LYSING BUFFER 100ML BOTTLE
SDP
Catalog No. 501018893
Encompass_Preferred
Ammonium-Chloride-Potassium is used to lyse red blood cells in preparations containing white blood cells. One 100ml bottle.
Catalog No. 50-101-8893 Supplier Kd Medical Inc Supplier No. RGC3015
Ammonium-Chloride-Potassium is used to lyse red blood cells in preparations containing white blood cells. One 100ml bottle.