Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ACKR3 protein, human (His-SUMO) | >95.0% | 10UG
SDP

Catalog No. 5000175465
Encompass_Preferred

Recombinant human ACKR3 extracellular domain produced in E. coli, supplied as a lyophilized His-SUMO-tagged protein for biochemical and binding studies.

  • Recombinant human ACKR3 extracellular domain with N-terminal His-SUMO tag.
  • Expressed in E. coli and supplied as lyophilized powder.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Approximate molecular weight 27 kDa.
  • Endotoxin level below 1 EU/μg.
  • Lyophilized formulation includes trehalose for stability.
  • Store frozen; reconstitute to ≥100 μg/mL for use.
  • Suitable for ligand binding, β-arrestin recruitment, and MAPK pathway assays.

Catalog No. 50-001-75465 Supplier Medchemexpress LLC Supplier No. HYP70053710UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human ACKR3 extracellular domain produced in E. coli, supplied as a lyophilized His-SUMO-tagged protein for biochemical and binding studies.

  • Recombinant human ACKR3 extracellular domain with N-terminal His-SUMO tag.
  • Expressed in E. coli and supplied as lyophilized powder.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Approximate molecular weight 27 kDa.
  • Endotoxin level below 1 EU/μg.
  • Lyophilized formulation includes trehalose for stability.
  • Store frozen; reconstitute to ≥100 μg/mL for use.
  • Suitable for ligand binding, β-arrestin recruitment, and MAPK pathway assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.